Methyl-2-(4-methylpiperazino)-5-nitrobenzenecarboxylate ≥95%
Supplier: Matrix Scientific
047968-500MG
047968-1G
047968-5G
101882-208EA
278.46
USD
101882-208
101882-210
101882-212
Methyl-2-(4-methylpiperazino)-5-nitrobenzenecarboxylate ≥95%
Methyl-2-(4-methylpiperazino)-5-nitrobenzenecarboxylate
Hazard: Irritant
Formula:
C₁₃H₁₇N₃O₄ Melting Pt: 110…112 °C |
MDL Number:
MFCD11553067 CAS Number: 735213-47-5 |
Specification Test Results
Melting point | 110-112°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...