4-Iodo-N,N-dimethyl-1H-pyrrole-2-carboxamide ≥95%
Supplier: Matrix Scientific
047967-500MG
047967-1G
101882-204EA
556.92
USD
101882-204
101882-206
4-Iodo-N,N-dimethyl-1H-pyrrole-2-carboxamide ≥95%
4-Iodo-N,N-dimethyl-1H-pyrrole-2-carboxamide
Hazard: Irritant
Formula:
C₇H₉IN₂O Melting Pt: 181…182 °C |
MDL Number:
MFCD11553066 CAS Number: 1092352-92-5 |
Specification Test Results
Melting point | 181-182°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...