(5-Bromo-2-thienyl)(piperazino)methanone ≥95%

Supplier: Matrix Scientific

047961-500MG 047961-1G
101882-178EA 280 USD
101882-178 101882-180
(5-Bromo-2-thienyl)(piperazino)methanone ≥95%
(5-Bromo-2-thienyl)(piperazino)methanone

Hazard: Irritant

Formula: C₉H₁₁BrN₂OS
Melting Pt: 86…87 °C
MDL Number: MFCD08310617
CAS Number: 395648-06-3

Order Now

Specification Test Results

Melting point 86-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR