4-(1,3-Thiazolan-2-yl)pyridine ≥95%

Supplier: Matrix Scientific

047960-500MG 047960-1G
101882-174EA 181.54 USD
101882-174 101882-176
4-(1,3-Thiazolan-2-yl)pyridine ≥95%
4-(1,3-Thiazolan-2-yl)pyridine

Hazard: Irritant

Formula: C₈H₁₀N₂S
Melting Pt: 82…83 °C
MDL Number: MFCD01681087
CAS Number: 700-92-5

Order Now

Specification Test Results

Melting point 82-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR