tert-Butyl 4-[(5-bromo-3-pyridinyl)carbonyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Supplier: Matrix Scientific
047959-500MG
047959-1G
047959-5G
101882-168EA
181.54
USD
101882-168
101882-170
101882-172
tert-Butyl 4-[(5-bromo-3-pyridinyl)carbonyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl 4-[(5-bromo-3-pyridinyl)carbonyl]tetrahydro-1(2H)-pyrazinecarboxylate
Hazard: Irritant
Formula:
C₁₅H₂₀BrN₃O₃ MW: 370.25 g/mol Melting Pt: 120…122 °C |
MDL Number:
MFCD09972241 CAS Number: 1017782-72-7 |
Specification Test Results
Melting point | 120-122°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...