tert-Butyl-4-[(5-bromo-2-thienyl)carbonyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

047958-500MG 047958-1G 047958-5G
101882-162EA 181.54 USD
101882-162 101882-164 101882-166
tert-Butyl-4-[(5-bromo-2-thienyl)carbonyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-[(5-bromo-2-thienyl)carbonyl]tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₉BrN₂O₃S
MW: 375.29 g/mol
Melting Pt: 107…108 °C
MDL Number: MFCD09972240
CAS Number: 1017782-71-6

Order Now

Specification Test Results

Melting point 107-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR