Benzyl-4-{[(2-bromoethyl)(tert-butoxycarbonyl)amino]sulfonyl}tetrahydro-1(2H)-pyrazine carboxylate ≥90%

Supplier: Matrix Scientific

047956-500MG 047956-1G
101882-154EA 375.38 USD
101882-154 101882-156
Benzyl-4-{[(2-bromoethyl)(tert-butoxycarbonyl)amino]sulfonyl}tetrahydro-1(2H)-pyrazine carboxylate ≥90%
Benzyl-4-{[(2-bromoethyl)(tert-butoxycarbonyl)amino]sulfonyl}tetrahydro-1(2H)-pyrazine carboxylate

Hazard: Irritant

Formula: C₁₉H₂₈BrN₃O₆S
Melting Pt: 125…126 °C
MDL Number: MFCD09972239
CAS Number: 1017782-70-5

Order Now

Specification Test Results

Melting point 125-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR