tert-Butyl-3-[4-(methoxycarbonyl)phenyl]-2,2-dioxo-2λ~6~-diazathiane-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
2,2-Dioxo-3-[4-(methoxycarbonyl)phenyl]diazathiane, N1-BOC protected
047954-500MG
047954-1G
047954-5G
101882-144EA
181.54
USD
101882-144
101882-146
101882-148
tert-Butyl-3-[4-(methoxycarbonyl)phenyl]-2,2-dioxo-2λ~6~-diazathiane-1-carboxylate ≥95%
tert-Butyl-3-[4-(methoxycarbonyl)phenyl]-2,2-dioxo-2lambda~6~-diazathiane-1-carboxylate
Hazard: Irritant
Formula:
C₁₃H₁₈N₂O₆S MW: 330.36 g/mol Melting Pt: 135…136 °C |
MDL Number:
MFCD09865024 CAS Number: 1017782-69-2 |
Specification Test Results
Melting point | 135-136°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...