tert-Butyl-2,2-dioxo-3-(1,3-thiazol-2-yl)-2λ~6~-diazathiane-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
2,2-Dioxo-3-(1,3-thiazol-2-yl)diazathiane, N1-BOC protected
047952-500MG
047952-1G
047952-5G
101882-132EA
181.54
USD
101882-132
101882-134
101882-136
tert-Butyl-2,2-dioxo-3-(1,3-thiazol-2-yl)-2λ~6~-diazathiane-1-carboxylate ≥95%
tert-Butyl-2,2-dioxo-3-(1,3-thiazol-2-yl)-2lambda~6~-diazathiane-1-carboxylate
Hazard: Irritant
Formula:
C₈H₁₃N₃O₄S₂ MW: 279.34 g/mol Melting Pt: 143…145 °C |
MDL Number:
MFCD09865022 CAS Number: 1017782-67-0 |
Specification Test Results
Melting point | 143-145°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...