tert-Butyl-3-(3-cyanophenyl)-2,2-dioxo-2λ~6~-diazathiane-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(3-Cyanophenyl)-2,2-dioxodiazathiane, N1-BOC protected

047951-500MG 047951-1G 047951-5G
101882-126EA 181.54 USD
101882-126 101882-128 101882-130
tert-Butyl-3-(3-cyanophenyl)-2,2-dioxo-2λ~6~-diazathiane-1-carboxylate ≥95%
tert-Butyl-3-(3-cyanophenyl)-2,2-dioxo-2lambda~6~-diazathiane-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₅N₃O₄S
MW: 297.34 g/mol
Melting Pt: 140…141 °C
MDL Number: MFCD09865021
CAS Number: 1017782-66-9

Order Now

Specification Test Results

Melting point 140-141°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR