5-Bromo-4-(4-fluorophenyl)-1,3-thiazol-2-amine hydrobromide ≥95%
Supplier: Matrix Scientific
047949-500MG
047949-1G
047949-5G
101882-114EA
284.62
USD
101882-114
101882-116
101882-118
5-Bromo-4-(4-fluorophenyl)-1,3-thiazol-2-amine hydrobromide ≥95%
5-Bromo-4-(4-fluorophenyl)-1,3-thiazol-2-amine hydrobromide
Hazard: Irritant
Formula:
C₉H₇Br₂FN₂S Melting Pt: 180…182 °C |
MDL Number:
MFCD09972237 CAS Number: 1171027-89-6 |
Specification Test Results
Melting point | 180-182°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...