4-(Dimethylamino)-3-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Dimethylamino)-3-nitrobenzaldehyde

047947-500MG 047947-1G 047947-5G
101882-108EA 166.15 USD
101882-108 101882-110 101882-112
4-(Dimethylamino)-3-nitrobenzenecarbaldehyde ≥95%
4-(Dimethylamino)-3-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₉H₁₀N₂O₃
MW: 194.19 g/mol
Melting Pt: 105…107 °C
MDL Number: MFCD00480831
CAS Number: 59935-39-6

Order Now

Specification Test Results

Melting point 105-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR