Methyl-3-amino-6-bromobenzo[b]thiophene-2-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 3-amino-6-bromobenzo[b]thiophene-2-carboxylate
047940-500MG
047940-1G
047940-5G
101882-082EA
178.46
USD
101882-082
101882-084
101882-086
Methyl-3-amino-6-bromobenzo[b]thiophene-2-carboxylate ≥95%
Methyl 3-amino-6-bromobenzo[b]thiophene-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₈BrNO₂S MW: 286.1450 g/mol Melting Pt: 126-128°C |
MDL Number:
MFCD09865017 CAS Number: 1017782-63-6 |
Specification Test Results
Melting point | 126-128°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...