tert-Butyl-4-[3-bromo-5-(methoxycarbonyl)-2-pyridinyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
4-[3-Bromo-5-(methoxycarbonyl)pyridin-2-yl]piperazine, N1-BOC protected
047936-500MG
047936-1G
047936-5G
101882-068EA
492.31
USD
101882-068
101882-070
101882-072
tert-Butyl-4-[3-bromo-5-(methoxycarbonyl)-2-pyridinyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-[3-bromo-5-(methoxycarbonyl)-2-pyridinyl]tetrahydro-1(2H)-pyrazinecarboxylate
Hazard: Irritant
Formula:
C₁₆H₂₂BrN₃O₄ MW: 400.28 g/mol Melting Pt: 90…92 °C |
MDL Number:
MFCD09865015 CAS Number: 906559-32-8 |
Specification Test Results
Melting point | 90-92°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...