4-{2-[4-(Trifluoromethyl)piperidino]acetyl}-1H-pyrrole-2-carbaldehyde ≥95%
Supplier: Matrix Scientific
047934-500MG
047934-1G
101882-058EA
375.38
USD
101882-058
101882-060
4-{2-[4-(Trifluoromethyl)piperidino]acetyl}-1H-pyrrole-2-carbaldehyde ≥95%
4-{2-[4-(Trifluoromethyl)piperidino]acetyl}-1H-pyrrole-2-carbaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₅F₃N₂O₂ MW: 288.27 g/mol Melting Pt: 142…144 °C |
MDL Number:
MFCD09864685 CAS Number: 1000339-90-1 |
Specification Test Results
Melting point | 142-144°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...