5-Bromo-4-(4-bromophenyl)-1,3-thiazol-2-amine hydrobromide ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-bromo-4-(4-bromophenyl)-1,3-thiazole hydrobromide

047933-500MG 047933-1G 047933-5G
101882-052EA 375.38 USD
101882-052 101882-054 101882-056
5-Bromo-4-(4-bromophenyl)-1,3-thiazol-2-amine hydrobromide ≥95%
5-Bromo-4-(4-bromophenyl)-1,3-thiazol-2-amine hydrobromide

Hazard: Irritant

Formula: C₉H₇Br₃N₂S
MW: 414.96 g/mol
Melting Pt: 223…225 °C
MDL Number: MFCD09865014
CAS Number: 1172108-91-6

Order Now

Specification Test Results

Melting point 223-225°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR