Methyl-3-amino-3-(1,3-benzodioxol-5-yl)propanoate hydrochloride ≥95%
Supplier: Matrix Scientific
047932-500MG
047932-1G
047932-5G
101882-046EA
278.46
USD
101882-046
101882-048
101882-050
Methyl-3-amino-3-(1,3-benzodioxol-5-yl)propanoate hydrochloride ≥95%
Methyl-3-amino-3-(1,3-benzodioxol-5-yl)propanoate hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₄ClNO₄ MW: 259.69 g/mol Melting Pt: 175…177 °C |
MDL Number:
MFCD09864841 CAS Number: 1177093-05-8 |
Specification Test Results
Melting point | 175-177°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...