2-Bromo-3-(4-bromophenyl)-5,6-dihydroimidazo[2,1-b][1,3]thiazole hydrobromide ≥95%
Supplier: Matrix Scientific
047925-500MG
047925-1G
047925-5G
101882-024EA
278.46
USD
101882-024
101882-026
101882-028
2-Bromo-3-(4-bromophenyl)-5,6-dihydroimidazo[2,1-b][1,3]thiazole hydrobromide ≥95%
2-Bromo-3-(4-bromophenyl)-5,6-dihydroimidazo[2,1-b][1,3]thiazole hydrobromide
Hazard: Irritant
Formula:
C₁₁H₉Br₃N₂S Melting Pt: >250 °C |
MDL Number:
MFCD09865013 CAS Number: 1172759-40-8 |
Specification Test Results
Melting point | >250°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...