5-(1,4-Thiazinan-4-yl)-2-thiophenecarbaldehyde ≥95%
Supplier: Matrix Scientific
047912-500MG
047912-1G
047912-5G
101881-968EA
278.46
USD
101881-968
101881-970
101881-972
5-(1,4-Thiazinan-4-yl)-2-thiophenecarbaldehyde ≥95%
5-(1,4-Thiazinan-4-yl)-2-thiophenecarbaldehyde
Hazard: Irritant
Formula:
C₉H₁₁NOS₂ Melting Pt: 108…110 °C |
MDL Number:
MFCD09027177 CAS Number: 937604-40-5 |
Specification Test Results
Melting point | 108-110°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...