2-Bromoethyl-4-(1,3-thiazolan-2-yl)phenyl ether ≥95%
Supplier: Matrix Scientific
047911-500MG
047911-1G
047911-5G
101881-962EA
375.38
USD
101881-962
101881-964
101881-966
2-Bromoethyl-4-(1,3-thiazolan-2-yl)phenyl ether ≥95%
2-Bromoethyl-4-(1,3-thiazolan-2-yl)phenyl ether
Hazard: Irritant
Formula:
C₁₁H₁₄BrNOS Melting Pt: 96…98 °C |
MDL Number:
MFCD09027176 CAS Number: 937604-18-7 |
Specification Test Results
Melting point | 96-98°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...