Methyl-6-bromobenzo[b]thiophene-2-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 6-bromo-1-benzothiophene-2-carboxylate, Methyl 6-bromobenzo[b]thiophene-2-carboxylate
047910-500MG
047910-1G
047910-5G
101881-956EA
92.31
USD
101881-956
101881-958
101881-960
Methyl-6-bromobenzo[b]thiophene-2-carboxylate ≥95%
Methyl 6-bromobenzo[b]thiophene-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₇BrO₂S MW: 271.1300 g/mol Melting Pt: 117-119°C |
MDL Number:
MFCD07371541 CAS Number: 360576-01-8 |
Specification Test Results
Melting point | 117-119°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...