3-Fluoropropyl-4-(1,3-thiazolan-2-yl)phenyl ether ≥95%

Supplier: Matrix Scientific

047909-500MG 047909-1G 047909-5G
101881-950EA 556.92 USD
101881-950 101881-952 101881-954
3-Fluoropropyl-4-(1,3-thiazolan-2-yl)phenyl ether ≥95%
3-Fluoropropyl-4-(1,3-thiazolan-2-yl)phenyl ether

Hazard: Irritant

Formula: C₁₂H₁₆FNOS
Melting Pt: 90…92 °C
MDL Number: MFCD09027175
CAS Number: 937604-32-5

Order Now

Specification Test Results

Melting point 90-92°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR