1-Benzyl-2-[4-(tert-butyl)phenyl]-6-oxo-3-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

047908-500MG 047908-1G 047908-5G
101881-944EA 181.54 USD
101881-944 101881-946 101881-948
1-Benzyl-2-[4-(tert-butyl)phenyl]-6-oxo-3-piperidinecarboxylic acid ≥95%
1-Benzyl-2-[4-(tert-butyl)phenyl]-6-oxo-3-piperidinecarboxylic acid
Formula: C₂₃H₂₇NO₃
Melting Pt: 144…146 °C
MDL Number: MFCD09027174
CAS Number: 952182-64-8

Order Now

Specification Test Results

Melting point 144-146°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR