2-(5-Chloro-2-phenoxyphenyl)acetic acid ≥95%

Supplier: Matrix Scientific

047905-500MG 047905-1G
101881-928EA 375.38 USD
101881-928 101881-930
2-(5-Chloro-2-phenoxyphenyl)acetic acid ≥95%
2-(5-Chloro-2-phenoxyphenyl)acetic acid

Hazard: Irritant

Formula: C₁₄H₁₁ClO₃
MW: 262.6900 g/mol
Melting Pt: 123…125 °C
MDL Number: MFCD07368830
CAS Number: 70958-20-2

Order Now

Specification Test Results

Melting point 123-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR