Amlodipine besylate ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(2-Aminoethoxy)methyl-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylic acid 3-Ethyl 5-Methyl Ester Benzenesulphon, 3-Ethyl 5-Methyl 2-(2-Aminoethoxy)methyl-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate Benzenesulphonate, 3-Ethyl 5-methyl 2-((2-aminoethoxy)methyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate benzenesulphonate

047879-1G 047879-5G
101881-824EA 307.69 USD
101881-824 102551-378
Amlodipine besylate ≥95%
Amlodipine besylate

Hazard: Irritant

Formula: C₂₀H₂₅ClN₂O₅
MW: 567.05 g/mol
Melting Pt: 198…200 °C
MDL Number: MFCD00887594
CAS Number: 111470-99-6

Order Now

Specification Test Results

Melting point 198-200°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR