1-Methyl-6-(2-pyridinyl)-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%

Supplier: Matrix Scientific

047850-500MG 047850-1G
101881-700EA 747.69 USD
101881-700 101881-702
1-Methyl-6-(2-pyridinyl)-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%
1-Methyl-6-(2-pyridinyl)-1H-pyrazolo[3,4-b]pyridin-3-amine

Hazard: Irritant

Formula: C₁₂H₁₁N₅
Melting Pt: 177…179 °C
MDL Number: MFCD11553063
CAS Number: 1092352-88-9

Order Now

Specification Test Results

Melting point 177-179°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR