7-Bromo-2-methyl-1,3-benzoxazol-5-yl methyl ether ≥95%

Supplier: Matrix Scientific

047842-500MG 047842-1G
101881-670EA 315.38 USD
101881-670 101881-672
7-Bromo-2-methyl-1,3-benzoxazol-5-yl methyl ether ≥95%
7-Bromo-2-methyl-1,3-benzoxazol-5-yl methyl ether

Hazard: Irritant

Formula: C₉H₈BrNO₂
Melting Pt: 105…107 °C
MDL Number: MFCD11553059
CAS Number: 1092352-82-3

Order Now

Specification Test Results

Melting point 105-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR