Methyl amino(3-bromophenyl)acetate hydrochloride ≥95%
Supplier: Matrix Scientific
047838-500MG
047838-1G
101881-658EA
483.08
USD
101881-658
101881-660
Methyl amino(3-bromophenyl)acetate hydrochloride ≥95%
Methyl amino(3-bromophenyl)acetate hydrochloride
Hazard: Irritant
Formula:
C₉H₁₁BrClNO₂ Melting Pt: 210 °C |
MDL Number:
MFCD11841010 CAS Number: 1219198-88-5 |
Specification Test Results
Melting point | 210°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...