4-[2-(Bromomethyl)-4-nitrophenyl]morpholine ≥95%

Supplier: Matrix Scientific

047836-500MG 047836-1G
101881-650EA 375.38 USD
101881-650 101881-652
4-[2-(Bromomethyl)-4-nitrophenyl]morpholine ≥95%
4-[2-(Bromomethyl)-4-nitrophenyl]morpholine

Hazard: Irritant

Formula: C₁₁H₁₃BrN₂O₃
Melting Pt: 85…88 °C
MDL Number: MFCD11553056
CAS Number: 1092352-78-7

Order Now

Specification Test Results

Melting point 85-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR