Methyl-3-amino-6-(2-pyridinyl)thieno[2,3-b]pyridine-2-carboxylate ≥95%
Supplier: Matrix Scientific
047831-500MG
047831-1G
101881-628EA
556.92
USD
101881-628
101881-630
Methyl-3-amino-6-(2-pyridinyl)thieno[2,3-b]pyridine-2-carboxylate ≥95%
Methyl-3-amino-6-(2-pyridinyl)thieno[2,3-b]pyridine-2-carboxylate
Hazard: Irritant
Formula:
C₁₄H₁₁N₃O₂S Melting Pt: 275…277 °C |
MDL Number:
MFCD08560632 CAS Number: 921135-52-6 |
Specification Test Results
Melting point | 275-277°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...