6-(2-Pyridinyl)-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%

Supplier: Matrix Scientific

047828-500MG 047828-1G
101881-614EA 556.92 USD
101881-614 101881-616
6-(2-Pyridinyl)-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%
6-(2-Pyridinyl)-1H-pyrazolo[3,4-b]pyridin-3-amine

Hazard: Irritant

Formula: C₁₁H₉N₅
Melting Pt: 239…241 °C
MDL Number: MFCD09027911
CAS Number: 946385-33-7

Order Now

Specification Test Results

Melting point 239-241°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR