5-Bromo-2-chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile ≥95%
Supplier: Matrix Scientific
047826-500MG
047826-1G
101881-608EA
353.85
USD
101881-608
101881-610
5-Bromo-2-chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile ≥95%
5-Bromo-2-chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile
Hazard: Irritant
Formula:
C₈H₃BrClF₃N₂ MW: 299.48 g/mol Melting Pt: 74…76 °C |
MDL Number:
MFCD11841008 CAS Number: 1135283-49-6 |
Specification Test Results
Melting point | 74-76°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...