Methyl-3-amino-5-bromo-6-(2-pyridinyl)thieno[2,3-b]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

047821-500MG 047821-1G
101881-592EA 921.54 USD
101881-592 101881-594
Methyl-3-amino-5-bromo-6-(2-pyridinyl)thieno[2,3-b]pyridine-2-carboxylate ≥95%
Methyl-3-amino-5-bromo-6-(2-pyridinyl)thieno[2,3-b]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₀BrN₃O₂S
Melting Pt: 224…226 °C
MDL Number: MFCD11553053
CAS Number: 1092352-72-1

Order Now

Specification Test Results

Melting point 224-226°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR