5-Nitro-1-benzofuran-2-carbohydrazide ≥95%

Supplier: Matrix Scientific

047820-500MG 047820-1G
101881-588EA 278.46 USD
101881-588 101881-590
5-Nitro-1-benzofuran-2-carbohydrazide ≥95%
5-Nitro-1-benzofuran-2-carbohydrazide

Hazard: Irritant

Formula: C₉H₇N₃O₄
MW: 221.17 g/mol
Melting Pt: 256…258 °C
MDL Number: MFCD11553052
CAS Number: 406192-63-0

Order Now

Specification Test Results

Melting point 256-258°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR