5-Amino-1-[2-nitro-4-(trifluoromethyl)phenyl]-1H-pyrazole-4-carbonitrile ≥95%
Supplier: Matrix Scientific
047816-500MG
047816-1G
047816-5G
101881-574EA
360
USD
101881-574
101881-576
101881-578
5-Amino-1-[2-nitro-4-(trifluoromethyl)phenyl]-1H-pyrazole-4-carbonitrile ≥95%
5-Amino-1-[2-nitro-4-(trifluoromethyl)phenyl]-1H-pyrazole-4-carbonitrile
Hazard: Irritant
Formula:
C₁₁H₆F₃N₅O₂ Melting Pt: 231…233 °C |
MDL Number:
MFCD00205968 CAS Number: 74990-43-5 |
Specification Test Results
Melting point | 231-233°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...