5-Chloro-2-hydroxy-6-methyl-4-(trifluoromethyl)nicotinonitrile ≥95%

Supplier: Matrix Scientific

047815-500MG 047815-1G
101881-570EA 278.46 USD
101881-570 101881-572
5-Chloro-2-hydroxy-6-methyl-4-(trifluoromethyl)nicotinonitrile ≥95%
5-Chloro-2-hydroxy-6-methyl-4-(trifluoromethyl)nicotinonitrile

Hazard: Irritant

Formula: C₈H₄ClF₃N₂O
MW: 236.58 g/mol
Melting Pt: 234…236 °C
MDL Number: MFCD11501054
CAS Number: 1092352-68-5

Order Now

Specification Test Results

Melting point 234-236°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR