Ethyl-1-{[(tert-butoxycarbonyl)amino]sulfonyl}-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Ethoxycarbonyl)piperidine-1-sulphonamide, N-BOC protected

047811-500MG 047811-1G 047811-5G
101881-546EA 104.62 USD
101881-546 101881-548 101881-550
Ethyl-1-{[(tert-butoxycarbonyl)amino]sulfonyl}-4-piperidinecarboxylate ≥95%
Ethyl-1-{[(tert-butoxycarbonyl)amino]sulfonyl}-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₃H₂₄N₂O₆S
MW: 336.41 g/mol
Melting Pt: 79…81 °C
MDL Number: MFCD09864838
CAS Number: 1000018-25-6

Order Now

Specification Test Results

Melting point 79-81°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR