2-Benzyl-1,2,7-thiadiazepane-1,1-dioxide ≥95%
Supplier: Matrix Scientific
047807-500MG
047807-1G
101881-530EA
146.15
USD
101881-530
101881-532
2-Benzyl-1,2,7-thiadiazepane-1,1-dioxide ≥95%
2-Benzyl-1,2,7-thiadiazepane-1,1-dioxide
Hazard: Irritant
Formula:
C₁₁H₁₆N₂O₂S Melting Pt: 72…74 °C |
MDL Number:
MFCD09972235 CAS Number: 1017782-61-4 |
Specification Test Results
Melting point | 72-74°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...