Ethyl-2-[6-oxo-4-piperazino-1(6H)-pyridazinyl]acetate ≥95%
Supplier: Matrix Scientific
047806-500MG
047806-1G
101881-526EA
556.92
USD
101881-526
101881-528
Ethyl-2-[6-oxo-4-piperazino-1(6H)-pyridazinyl]acetate ≥95%
Ethyl-2-[6-oxo-4-piperazino-1(6H)-pyridazinyl]acetate
Hazard: Irritant
Formula:
C₁₂H₁₈N₄O₃ MW: 266.3 g/mol Melting Pt: 145…150 °C |
MDL Number:
MFCD09864837 CAS Number: 1000018-24-5 |
Specification Test Results
Melting point | 145-150°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...