Benzyl-4-{[(tert-butoxycarbonyl)amino]sulfonyl}tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Piperazine-1-sulphonamide, N1-BOC N4-CBZ protected

047805-500MG 047805-1G 047805-5G
101881-520EA 181.54 USD
101881-520 101881-522 101881-524
Benzyl-4-{[(tert-butoxycarbonyl)amino]sulfonyl}tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Benzyl-4-{[(tert-butoxycarbonyl)amino]sulfonyl}tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₇H₂₅N₃O₆S
MW: 399.47 g/mol
Melting Pt: 98…110 °C
MDL Number: MFCD09864831
CAS Number: 1000018-21-2

Order Now

Specification Test Results

Melting point 98-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR