2-[4-{4-[(Benzyloxy)carbonyl]piperazino}-5-chloro-6-oxo-1(6H)-pyridazinyl]acetic acid ≥95%
Supplier: Matrix Scientific
047804-500MG
047804-1G
047804-5G
101881-514EA
375.38
USD
101881-514
101881-516
101881-518
2-[4-{4-[(Benzyloxy)carbonyl]piperazino}-5-chloro-6-oxo-1(6H)-pyridazinyl]acetic acid ≥95%
2-[4-{4-[(Benzyloxy)carbonyl]piperazino}-5-chloro-6-oxo-1(6H)-pyridazinyl]acetic acid
Hazard: Irritant
Formula:
C₁₈H₁₉ClN₄O₅ Melting Pt: 129…132 °C |
MDL Number:
MFCD09864830 CAS Number: 1000018-20-1 |
Specification Test Results
Melting point | 129-132°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...