Methyl-4-{2-[4-(trifluoromethyl)piperidino]ethoxy}benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-{2-[4-(trifluoromethyl)piperidin-1-yl]ethoxy}benzoate

047800-500MG 047800-1G
101881-498EA 375.38 USD
101881-498 101881-500
Methyl-4-{2-[4-(trifluoromethyl)piperidino]ethoxy}benzenecarboxylate ≥95%
Methyl-4-{2-[4-(trifluoromethyl)piperidino]ethoxy}benzenecarboxylate

Hazard: Irritant

Formula: C₁₆H₂₀F₃NO₃
MW: 331.34 g/mol
Melting Pt: 79…81 °C
MDL Number: MFCD09864682
CAS Number: 1000339-87-6

Order Now

Specification Test Results

Melting point 79-81°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR