1-[4-(4-Bromophenyl)-1,3-thiazol-2-yl]-4-piperidinecarboxylic acid ≥95%
Supplier: Matrix Scientific
047792-500MG
047792-1G
047792-5G
101881-456EA
556.92
USD
101881-456
101881-458
101881-460
1-[4-(4-Bromophenyl)-1,3-thiazol-2-yl]-4-piperidinecarboxylic acid ≥95%
1-[4-(4-Bromophenyl)-1,3-thiazol-2-yl]-4-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₅H₁₅BrN₂O₂S MW: 367.27 g/mol Melting Pt: 192…194 °C |
MDL Number:
MFCD01990266 CAS Number: 296899-02-0 |
Specification Test Results
Melting point | 192-194°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...