2-[4-(Trifluoromethyl)piperidino]-1,3-thiazole-5-carbaldehyde ≥95%
Supplier: Matrix Scientific
047789-500MG
047789-1G
101881-442EA
921.54
USD
101881-442
101881-444
2-[4-(Trifluoromethyl)piperidino]-1,3-thiazole-5-carbaldehyde ≥95%
2-[4-(Trifluoromethyl)piperidino]-1,3-thiazole-5-carbaldehyde
Hazard: Irritant
Formula:
C₁₀H₁₁F₃N₂OS MW: 264.27 g/mol Melting Pt: 42…45 °C |
MDL Number:
MFCD09864683 CAS Number: 1000339-88-7 |
Specification Test Results
Melting point | 42-45°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...