(5-Bromo-1-methyl-1H-imidazol-2-yl)methanol ≥97%
Supplier: Matrix Scientific
047786-500MG
047786-1G
101881-432EA
261.54
USD
101881-432
101881-434
(5-Bromo-1-methyl-1H-imidazol-2-yl)methanol ≥97%
(5-Bromo-1-methyl-1H-imidazol-2-yl)methanol
Hazard: Irritant
Formula:
C₅H₇BrN₂O Melting Pt: 143…145 °C |
MDL Number:
MFCD09864833 CAS Number: 861362-06-3 |
Specification Test Results
Melting point | 143-145°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...