2-(Chloromethyl)-1-methyl-1H-imidazole ≥95%

Supplier: Matrix Scientific

047785-250MG 047785-1G
101881-428EA 244.7 USD
101881-428 101881-430
2-(Chloromethyl)-1-methyl-1H-imidazole ≥95%
2-(Chloromethyl)-1-methyl-1H-imidazole

Hazard: Irritant

Formula: C₅H₇ClN₂
Melting Pt: 182…184 °C
MDL Number: MFCD00234119
CAS Number: 19225-92-4

Order Now

Specification Test Results

Melting point 182-184°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR