2-Bromo-6-nitro-1,3-benzothiazole ≥95%

Supplier: Matrix Scientific

047783-500MG 047783-1G 047783-5G
101881-416EA 338.46 USD
101881-416 101881-418 101881-420
2-Bromo-6-nitro-1,3-benzothiazole ≥95%
2-Bromo-6-nitro-1,3-benzothiazole

Hazard: Irritant

Formula: C₇H₃BrN₂O₂S
Melting Pt: 206…207 °C
MDL Number: MFCD07783787
CAS Number: 2516-37-2

Order Now

Specification Test Results

Melting point 206-207°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR