Ethyl-5-bromo-1-methyl-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

047781-1G 047781-5G
101881-412EA 407.69 USD
101881-412 101881-414
Ethyl-5-bromo-1-methyl-1H-indole-2-carboxylate ≥95%
Ethyl-5-bromo-1-methyl-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₂BrNO₂
MW: 282.14 g/mol
Melting Pt: 83…85 °C
MDL Number: MFCD09027165
CAS Number: 91844-20-1

Order Now

Specification Test Results

Melting point 83-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR