5,6-Dihydro-4H-cyclopenta[b]thiophene-2-carbonylchloride ≥95%

Supplier: Matrix Scientific

047779-500MG 047779-1G 047779-5G
101881-398EA 181.54 USD
101881-398 101881-400 101881-402
5,6-Dihydro-4H-cyclopenta[b]thiophene-2-carbonylchloride ≥95%

Hazard: Irritant/Store in Freezer

Formula: C₈H₇ClOS
Melting Pt: 40…42 °C
MDL Number: MFCD08059499
CAS Number: 142329-25-7

Order Now

Specification Test Results

Melting point 40-42°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR