2-[4-(Methylsulfanyl)phenyl]tetrahydro-1H-1λ-6-isothiazole-1,1-dione ≥95%

Supplier: Matrix Scientific

047778-500MG 047778-1G 047778-5G
101881-392EA 375.38 USD
101881-392 101881-394 101881-396
2-[4-(Methylsulfanyl)phenyl]tetrahydro-1H-1λ-6-isothiazole-1,1-dione ≥95%
2-[4-(Methylsulfanyl)phenyl]tetrahydro-1H-1lambda-6-isothiazole-1,1-dione

Hazard: Irritant

Formula: C₁₀H₁₃NO₂S₂
MW: 243.35 g/mol
Melting Pt: 108…110 °C
MDL Number: MFCD09152741
CAS Number: 952182-63-7

Order Now

Specification Test Results

Melting point 108-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR